Assay Details
| Gene Symbol: nedd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034186.1 | NP_001029358.1 | 2628 | 130 | ||
| GenBank mRNA | AY995154.1 | - | 2628 | 130 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nedd4a| Gene Symbol: nedd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034186.1 | NP_001029358.1 | 2628 | 130 | ||
| GenBank mRNA | AY995154.1 | - | 2628 | 130 | ||
Set Membership: |
|