Assay Details
| Gene Symbol: slit1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034968.1 | NP_001030140.1 | 29 - 30 | 3228 | 75 | |
| XM_009307022.3 | XP_009305297.1 | 26 - 27 | 2947 | 75 | ||
| GenBank mRNA | DQ177286.1 | - | 29 - 30 | 3228 | 75 | |
More Information
Set Membership: |
|

slit1a
Genomic Map