Assay Details
| Gene Symbol: slit1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034968.1 | NP_001030140.1 | 4 - 5 | 535 | 75 | |
| GenBank mRNA | BC163538.1 | - | 4 - 5 | 427 | 75 | |
| DQ177286.1 | - | 4 - 5 | 535 | 75 | ||
More Information
Set Membership: |
|

slit1a
Genomic Map