Assay Details
| Gene Symbol: prkacbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034976.1 | NP_001030148.1 | 9 - 10 | 934 | 69 | |
| XM_005171094.4 | XP_005171151.1 | 10 - 11 | 1016 | 69 | ||
| GenBank mRNA | BC103487.1 | - | 9 - 10 | 934 | 69 | |
More Information
Set Membership: |
|

prkacbb
Genomic Map