Assay Details
| Gene Symbol: map2k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037391.2 | NP_001032468.2 | 2 - 3 | 801 | 64 | |
| GenBank mRNA | BC107821.1 | - | 2 - 3 | 810 | 64 | |
| EF433291.1 | - | 2 - 3 | 801 | 64 | ||
More Information
Set Membership: |
|

map2k2a
Genomic Map