Assay Details
| Gene Symbol: dcdc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037689.2 | NP_001032778.2 | 10 - 11 | 1070 | 85 | |
| XM_017358759.2 | XP_017214248.1 | 12 - 13 | 1457 | 85 | ||
| XM_017358760.2 | XP_017214249.1 | 12 - 13 | 1431 | 85 | ||
| XM_017358761.2 | XP_017214250.1 | 12 - 13 | 1444 | 85 | ||
| XM_017358762.2 | XP_017214251.1 | 12 - 13 | 1418 | 85 | ||
| XM_017358763.2 | XP_017214252.1 | 12 - 13 | 1436 | 85 | ||
| XM_017358764.2 | XP_017214253.1 | 12 - 13 | 1397 | 85 | ||
| XM_017358765.2 | XP_017214254.1 | 12 - 13 | 1457 | 85 | ||
| XM_021480609.1 | XP_021336284.1 | 11 - 12 | 2831 | 85 | ||
| GenBank mRNA | BC108062.1 | - | 10 - 11 | 1081 | 85 | |
More Information
Set Membership: |
|

dcdc2b
Genomic Map