Assay Details
| Gene Symbol: smtla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037706.1 | NP_001032795.1 | 3 - 4 | 374 | 65 | |
| XM_021467699.1 | XP_021323374.1 | 3 - 4 | 365 | 65 | ||
| GenBank mRNA | AY376857.1 | - | 3 - 4 | 374 | 65 | |
More Information
Set Membership: |
|

smtla
Genomic Map