Assay Details
Gene Symbol: si:dkey-102c8.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044345.1 | NP_001037810.1 | 1 - 2 | 294 | 91 | |
XM_005173958.4 | XP_005174015.1 | 2 - 3 | 239 | 91 | ||
XM_009296103.3 | XP_009294378.1 | 2 - 3 | 254 | 91 | ||
GenBank mRNA | BC135063.1 | - | 1 - 2 | 294 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
