Assay Details
| Gene Symbol: taf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044395.2 | NP_001037860.2 | 5 - 6 | 588 | 65 | |
| GenBank mRNA | BC076279.1 | - | 5 - 6 | 508 | 65 | |
| BC095147.1 | - | 5 - 6 | 497 | 65 | ||
| BC122276.1 | - | 5 - 6 | 555 | 65 | ||
| BC153648.1 | - | 5 - 6 | 508 | 65 | ||
| BC165107.1 | - | 5 - 6 | 505 | 65 | ||
| EH498335.1 | - | 5 - 6 | 617 | 65 | ||
| EH555285.1 | - | 5 - 6 | 560 | 65 | ||
More Information
Set Membership: |
|

taf9
Genomic Map