Assay Details
| Gene Symbol: poc1bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044778.2 | NP_001038243.2 | 2 - 3 | 682 | 66 | |
| GenBank mRNA | BC090692.1 | - | 2 - 3 | 682 | 66 | |
| BC164662.1 | - | 1 - 2 | 419 | 66 | ||
More Information
Set Membership: |
|

poc1bl
Genomic Map