Assay Details
| Gene Symbol: poc1bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044778.2 | NP_001038243.2 | 9 - 10 | 1761 | 118 | |
| GenBank mRNA | BC090692.1 | - | 9 - 10 | 1761 | 118 | |
| BC164662.1 | - | 8 - 9 | 1498 | 118 | ||
More Information
Set Membership: |
|

poc1bl
Genomic Map