Assay Details
| Gene Symbol: smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044795.1 | NP_001038260.1 | 9 - 10 | 1134 | 68 | |
| XM_005159676.4 | XP_005159733.1 | 9 - 10 | 1231 | 68 | ||
| GenBank mRNA | BC171527.1 | - | 9 - 10 | 1234 | 68 | |
| BC171531.1 | - | 9 - 10 | 1234 | 68 | ||
More Information
Set Membership: |
|

smap2
Genomic Map