Assay Details
| Gene Symbol: mapk12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044841.2 | NP_001038306.1 | 3 - 4 | 476 | 91 | |
| GenBank mRNA | BC133969.1 | - | 3 - 4 | 476 | 91 | |
More Information
Set Membership: |
|

mapk12b
Genomic Map