Assay Details
| Gene Symbol: synm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044875.2 | NP_001038340.1 | 3 - 4 | 1084 | 124 | |
| XM_021467662.1 | XP_021323337.1 | 2 - 3 | 322 | 124 | ||
| GenBank mRNA | BC171396.1 | - | 3 - 4 | 1084 | 124 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021467662.1 transcripts.
Set Membership: |
|

synm
Genomic Map