Assay Details
| Gene Symbol: eif3eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC057516.1 | - | 13 - 14 | 1414 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif3eb| Gene Symbol: eif3eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC057516.1 | - | 13 - 14 | 1414 | 69 | |
Set Membership: |
|