Assay Details
| Gene Symbol: eif3eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362621.1 | NP_001349550.1 | 9 - 10 | 985 | 77 | |
| GenBank mRNA | BC057516.1 | - | 9 - 10 | 964 | 77 | |
More Information
Set Membership: |
|

eif3eb
Genomic Map