Assay Details
| Gene Symbol: mtmr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045070.1 | NP_001038535.1 | 6 - 7 | 972 | 62 | |
| XM_005160464.4 | XP_005160521.1 | 7 - 8 | 1144 | 62 | ||
| GenBank mRNA | BC163574.1 | - | 6 - 7 | 1001 | 62 | |
| BC163591.1 | - | 6 - 7 | 1001 | 62 | ||
More Information
Set Membership: |
|

mtmr9
Genomic Map