Assay Details
| Gene Symbol: tab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045105.1 | NP_001038570.1 | 3 - 4 | 1481 | 79 | |
| XM_017357352.2 | XP_017212841.1 | 3 - 4 | 1789 | 79 | ||
| XM_021478287.1 | XP_021333962.1 | 3 - 4 | 1530 | 79 | ||
| GenBank mRNA | BC134845.1 | - | 3 - 4 | 1392 | 79 | |
More Information
Set Membership: |
|

tab3
Genomic Map