Assay Details
| Gene Symbol: fgf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045324.1 | NP_001038789.1 | 126 | 76 | ||
| GenBank mRNA | AB209932.1 | - | 126 | 76 | ||
| BC162750.1 | - | 155 | 76 | |||
More Information
Set Membership: |
|

fgf21
Genomic Map