Assay Details
| Gene Symbol: ncl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076652.2 | NP_001070120.2 | 7 - 8 | 901 | 152 | |
| XM_005170547.4 | XP_005170604.1 | 7 - 8 | 899 | 152 | ||
| XM_005170548.4 | XP_005170605.1 | 6 - 7 | 758 | 152 | ||
| GenBank mRNA | BC124135.1 | - | 7 - 8 | 848 | 152 | |
More Information
Set Membership: |
|

ncl
Genomic Map