Assay Details
| Gene Symbol: dennd2da | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077294.1 | NP_001070762.1 | 7 - 8 | 894 | 80 | |
| XM_017357375.2 | XP_017212864.1 | 10 - 11 | 1160 | 80 | ||
| GenBank mRNA | BC124709.1 | - | 7 - 8 | 894 | 80 | |
More Information
Set Membership: |
|

dennd2da
Genomic Map