Assay Details
| Gene Symbol: atp2a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077533.1 | NP_001071001.1 | 15 - 16 | 2002 | 64 | |
| XM_005156072.4 | XP_005156129.1 | 15 - 16 | 2036 | 64 | ||
| GenBank mRNA | BC051779.1 | - | 16 - 17 | 1886 | 64 | |
| BC125955.1 | - | 15 - 16 | 2002 | 64 | ||
More Information
Set Membership: |
|

atp2a1l
Genomic Map