Assay Details
| Gene Symbol: atp2a1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077533.1 | NP_001071001.1 | 18 - 19 | 2652 | 66 | |
| XM_005156072.4 | XP_005156129.1 | 18 - 19 | 2686 | 66 | ||
| GenBank mRNA | BC051779.1 | - | 19 - 20 | 2321 | 66 | |
| BC125955.1 | - | 18 - 19 | 2652 | 66 | ||
More Information
Set Membership: |
|

atp2a1l
Genomic Map