Assay Details
| Gene Symbol: tat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077554.1 | NP_001071022.1 | 5 - 6 | 706 | 63 | |
| GenBank mRNA | BC125947.1 | - | 5 - 6 | 706 | 63 | |
| BC165579.1 | - | 4 - 5 | 568 | 63 | ||
More Information
Set Membership: |
|

tat
Genomic Map