Assay Details
| Gene Symbol: znf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077610.1 | NP_001071078.1 | 10 - 11 | 1171 | 52 | |
| XM_009295759.2 | XP_009294034.1 | 10 - 11 | 1676 | 52 | ||
| XM_009295760.3 | XP_009294035.1 | 10 - 11 | 1980 | 52 | ||
| GenBank mRNA | BC124408.1 | - | 10 - 11 | 1171 | 52 | |
| BC165391.1 | - | 9 - 10 | 1173 | 52 | ||
More Information
Set Membership: |
|

znf76
Genomic Map