Assay Details
| Gene Symbol: mxra8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079960.3 | NP_001073429.3 | 4 - 5 | 557 | 89 | |
| GenBank mRNA | BC124787.1 | - | 4 - 5 | 526 | 89 | |
| CU638746.1 | - | 4 - 5 | 499 | 89 | ||
| EH585789.1 | - | 4 - 5 | 583 | 89 | ||
More Information
Set Membership: |
|

mxra8b
Genomic Map