Assay Details
| Gene Symbol: mxra8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079960.3 | NP_001073429.3 | 6 - 7 | 1202 | 70 | |
| GenBank mRNA | BC124787.1 | - | 6 - 7 | 1171 | 70 | |
More Information
Set Membership: |
|

mxra8b
Genomic Map