Assay Details
| Gene Symbol: mxra8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079960.3 | NP_001073429.3 | 7 - 8 | 1229 | 65 | |
| GenBank mRNA | BC124787.1 | - | 7 - 8 | 1198 | 65 | |
More Information
Set Membership: |
|

mxra8b
Genomic Map