Assay Details
| Gene Symbol: mxra8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079960.3 | NP_001073429.3 | 8 - 9 | 1298 | 82 | |
| GenBank mRNA | BC124787.1 | - | 8 - 9 | 1267 | 82 | |
| CU638746.1 | - | 8 - 9 | 1240 | 82 | ||
More Information
Set Membership: |
|

mxra8b
Genomic Map