Assay Details
| Gene Symbol: slc2a11l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080016.1 | NP_001073485.1 | 9 - 10 | 1091 | 124 | |
| GenBank mRNA | BC128857.1 | - | 9 - 10 | 1114 | 124 | |
More Information
Set Membership: |
|

slc2a11l
Genomic Map