Assay Details
| Gene Symbol: tecpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009306680.3 | XP_009304955.1 | 5 - 6 | 691 | 71 | |
| XM_021480288.1 | XP_021335963.1 | 5 - 6 | 817 | 71 | ||
| XM_021480289.1 | XP_021335964.1 | 5 - 6 | 815 | 71 | ||
| GenBank mRNA | BC129434.1 | - | 5 - 6 | 778 | 71 | |
| BC171562.1 | - | 5 - 6 | 697 | 71 | ||
| BC171564.1 | - | 5 - 6 | 697 | 71 | ||
More Information
Set Membership: |
|

tecpr1a
Genomic Map