Assay Details
| Gene Symbol: ranbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080571.2 | NP_001074040.1 | 1 - 2 | 562 | 73 | |
| GenBank mRNA | BC129352.1 | - | 1 - 2 | 563 | 73 | |
| BC165240.1 | - | 1 - 2 | 252 | 73 | ||
More Information
Set Membership: |
|

ranbp9
Genomic Map