Assay Details
| Gene Symbol: wasf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080590.1 | NP_001074059.1 | 6 - 7 | 805 | 73 | |
| GenBank mRNA | BC129169.1 | - | 6 - 7 | 805 | 73 | |
More Information
Set Membership: |
|

wasf3b
Genomic Map