Assay Details
| Gene Symbol: entpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083581.1 | NP_001077050.1 | 9 - 10 | 1357 | 72 | |
| XM_005157870.2 | XP_005157927.1 | 9 - 10 | 1444 | 72 | ||
| GenBank mRNA | BC134912.1 | - | 9 - 10 | 1357 | 72 | |
| EF446129.1 | - | 9 - 10 | 1362 | 72 | ||
More Information
Set Membership: |
|

entpd3
Genomic Map