Assay Details
| Gene Symbol: stxbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089376.1 | NP_001082845.1 | 7 - 8 | 618 | 63 | |
| GenBank mRNA | BC134211.1 | - | 7 - 8 | 611 | 63 | |
| BC171526.1 | - | 7 - 8 | 600 | 63 | ||
| BC171552.1 | - | 7 - 8 | 600 | 63 | ||
More Information
Set Membership: |
|

stxbp1b
Genomic Map