Assay Details
| Gene Symbol: gnrhr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098193.1 | NP_001091663.1 | 1 - 2 | 587 | 70 | |
| XM_005159164.4 | XP_005159221.1 | 2 - 3 | 70 | |||
| GenBank mRNA | EF534376.1 | - | 1 - 2 | 587 | 70 | |
| EF571595.1 | - | 1 - 2 | 70 | |||
More Information
Set Membership: |
|

gnrhr4
Genomic Map