Assay Details
| Gene Symbol: sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098250.1 | NP_001091720.1 | 1 - 2 | 159 | 78 | |
| XM_009298036.3 | XP_009296311.1 | 1 - 2 | 197 | 78 | ||
| GenBank mRNA | BC129175.1 | - | 1 - 2 | 160 | 78 | |
| CU458918.1 | - | 1 - 2 | 159 | 78 | ||
| JF714479.1 | - | 1 - 2 | 64 | 78 | ||
| KJ574205.1 | - | 1 - 2 | 148 | 78 | ||
More Information
Set Membership: |
|

sigirr
Genomic Map