Assay Details
| Gene Symbol: stoml3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098750.1 | NP_001092220.1 | 6 - 7 | 720 | 89 | |
| GenBank mRNA | BC142865.1 | - | 6 - 7 | 720 | 89 | |
More Information
Set Membership: |
|

stoml3a
Genomic Map