Assay Details
| Gene Symbol: hmgb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099251.2 | NP_001092721.2 | 4 - 5 | 591 | 60 | |
| GenBank mRNA | BC146744.1 | - | 4 - 5 | 491 | 60 | |
| EH575546.1 | - | 1 - 2 | 582 | 60 | ||
| EH581927.1 | - | 4 - 5 | 618 | 60 | ||
More Information
Set Membership: |
|

hmgb1b
Genomic Map