Assay Details
| Gene Symbol: plppr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099258.1 | NP_001092728.1 | 1 - 2 | 362 | 103 | |
| XM_021470095.1 | XP_021325770.1 | 1 - 2 | 613 | 103 | ||
| XM_021470096.1 | XP_021325771.1 | 3 - 4 | 768 | 103 | ||
| GenBank mRNA | BC142843.1 | - | 1 - 2 | 362 | 103 | |
More Information
Set Membership: |
|

plppr5b
Genomic Map