Assay Details
| Gene Symbol: plppr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099258.1 | NP_001092728.1 | 5 - 6 | 1064 | 85 | |
| XM_021470096.1 | XP_021325771.1 | 7 - 8 | 1470 | 85 | ||
| GenBank mRNA | BC142843.1 | - | 5 - 6 | 1064 | 85 | |
More Information
Set Membership: |
|

plppr5b
Genomic Map