Assay Details
| Gene Symbol: man1b1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099979.1 | NP_001093449.1 | 13 - 14 | 2043 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

man1b1b| Gene Symbol: man1b1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099979.1 | NP_001093449.1 | 13 - 14 | 2043 | 81 | |
Set Membership: |
|