Assay Details
| Gene Symbol: mybphb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100137.1 | NP_001093607.1 | 11 - 12 | 1661 | 98 | |
| GenBank mRNA | BC146703.1 | - | 11 - 12 | 1661 | 98 | |
| BC154250.1 | - | 11 - 12 | 1520 | 98 | ||
More Information
Set Membership: |
|

mybphb
Genomic Map