Assay Details
| Gene Symbol: anxa2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105600.1 | NP_001099070.1 | 1 - 2 | 58 | 70 | |
| GenBank mRNA | AY178797.1 | - | 2 - 3 | 74 | 70 | |
| BC076036.1 | - | 1 - 2 | 63 | 70 | ||
| BC110093.1 | - | 2 - 3 | 88 | 70 | ||
| BC153582.1 | - | 1 - 2 | 58 | 70 | ||
| BC162558.1 | - | 2 - 3 | 74 | 70 | ||
More Information
Set Membership: |
|

anxa2b
Genomic Map