Assay Details
| Gene Symbol: plcd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109700.1 | NP_001103170.1 | 3 - 4 | 542 | 109 | |
| XM_005162701.4 | XP_005162758.1 | 3 - 4 | 616 | 109 | ||
| GenBank mRNA | BC152502.1 | - | 3 - 4 | 542 | 109 | |
More Information
Set Membership: |
|

plcd1a
Genomic Map