Assay Details
| Gene Symbol: slc1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109703.1 | NP_001103173.1 | 6 - 7 | 1020 | 83 | |
| XM_009305801.3 | XP_009304076.1 | 6 - 7 | 1423 | 83 | ||
| GenBank mRNA | BC151992.1 | - | 6 - 7 | 1020 | 83 | |
| HM138697.1 | - | 5 - 6 | 928 | 83 | ||
More Information
Set Membership: |
|

slc1a6
Genomic Map