Assay Details
| Gene Symbol: tead3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109724.1 | NP_001103194.1 | 5 - 6 | 615 | 95 | |
| XM_005166144.4 | XP_005166201.1 | 5 - 6 | 616 | 95 | ||
| GenBank mRNA | BC151979.1 | - | 5 - 6 | 615 | 95 | |
More Information
Set Membership: |
|

tead3b
Genomic Map