Assay Details
| Gene Symbol: tead3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109724.1 | NP_001103194.1 | 6 - 7 | 752 | 78 | |
| XM_005166141.4 | XP_005166198.1 | 7 - 8 | 765 | 78 | ||
| XM_005166142.4 | XP_005166199.1 | 7 - 8 | 765 | 78 | ||
| XM_005166143.4 | XP_005166200.1 | 6 - 7 | 753 | 78 | ||
| XM_005166144.4 | XP_005166201.1 | 6 - 7 | 753 | 78 | ||
| XM_005166145.4 | XP_005166202.1 | 7 - 8 | 765 | 78 | ||
| XM_005166146.4 | XP_005166203.1 | 6 - 7 | 753 | 78 | ||
| GenBank mRNA | BC151979.1 | - | 6 - 7 | 752 | 78 | |
More Information
Set Membership: |
|

tead3b
Genomic Map