Assay Details
| Gene Symbol: klf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130936.1 | NP_571011.1 | 2 - 3 | 1110 | 73 | |
| GenBank mRNA | AF392996.1 | - | 2 - 3 | 1110 | 73 | |
| BC162384.1 | - | 2 - 3 | 940 | 73 | ||
| BC162393.1 | - | 2 - 3 | 940 | 73 | ||
More Information
Set Membership: |
|

klf1
Genomic Map