Assay Details
| Gene Symbol: ek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131095.1 | NP_571170.1 | 14 - 15 | 2641 | 90 | |
| GenBank mRNA | AJ005028.1 | - | 8 - 9 | 1043 | 90 | |
| U89295.1 | - | 14 - 15 | 2641 | 90 | ||
More Information
Set Membership: |
|

ek1
Genomic Map